missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl phenyl carbonate, 97%

Product Code. 15487368
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487368 100 g 100g
15477368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487368 Supplier Thermo Scientific Alfa Aesar Supplier No. H60867.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6627-89-0 | C11H14O3 | 194.23 g/mol

tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6627-89-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008804
InChI Key UXWVQHXKKOGTSY-UHFFFAOYSA-N
Synonym t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
PubChem CID 81113
IUPAC Name tert-butyl phenyl carbonate
SMILES CC(C)(C)OC(=O)OC1=CC=CC=C1

Specifications

Density 1.05
Boiling Point 83°C
Flash Point 101°C (214°F)
Linear Formula (CH3)3COCOOC6H5
Quantity 100 g
Beilstein 2361602
Solubility Information Slightly soluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material tert-Butyl phenyl carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.