missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl phenyl carbonate, 98%

Product Code. 11955151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11955151 25 g Glass bottle 25g
11965151 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11955151 Supplier Thermo Scientific Acros Supplier No. 154390250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6627-89-0 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6627-89-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008804
InChI Key UXWVQHXKKOGTSY-UHFFFAOYSA-N
Synonym t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
PubChem CID 81113
IUPAC Name tert-butyl phenyl carbonate
SMILES CC(C)(C)OC(=O)OC1=CC=CC=C1

Specifications

Color Colorless
Density 1.0500g/mL
Boiling Point 83.0°C (0.6 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OCO2C(CH3)3
Refractive Index 1.4805
Quantity 25 g
Fieser 05,86
Specific Gravity 1.05
Formula Weight 194.23
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material tert-Butyl phenyl carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.