missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene, 99%

Product Code. 10204040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10204040 500 mL Glass bottle 500mL
10346170 1 L Glass bottle 1 L
10311564 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204040 Supplier Thermo Scientific Acros Supplier No. 107875000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-06-6 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Melting Point -58.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 169.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C(CH3)3
Refractive Index 1.492
Quantity 500 mL
Beilstein 05, 415
Merck Index 15, 1553
Specific Gravity 0.86
Solubility Information Solubility in water: 0.03g/L water (20°C). Other solubilities: miscible with alcohol,ether and benzene
Viscosity 1.15 mPa.s (20°C)
Formula Weight 134.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.