missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylchlorosilane, 97%

Product Code. 11400663
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11400663

Brand: Thermo Scientific Alfa Aesar A12721.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58479-61-1 | C16H19ClSi | 274.863 g/mol

tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

Solubility
Miscible with chloroform and ethyl acetate. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.863
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl

Specifications

Boiling Point 90°C to 92°C (0.01mmHg)
Quantity 50 g
UN Number UN2987
Beilstein 644023
Flash Point 112°C (233°F)
Solubility Information Miscible with chloroform and ethyl acetate. Immiscible with water.
Formula Weight 274.87
Percent Purity 97%
Refractive Index 1.568
Sensitivity Moisture sensitive
Density 1.072
Chemical Name or Material tert-Butyldiphenylchlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.