missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone, TRC

Product Code. 30137998
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137998 25 g 25g
30137593 100 g 100g
30011219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137998 Supplier Toronto Research Chemicals Supplier No. TRCB69295025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10 H14 O2
Molecular Weight (g/mol) 166.22
Synonym Hydroquinone, tert-butyl- (6CI,7CI,8CI), 2-(1,1-Dimethylethyl)-1,4-benzenediol, 2-(1,1-Dimethylethyl)hydroquinone, 2-(tert-Butyl)-p-hydroquinone, 2-t-Butylhydroquinone, 2-tert-Butyl-1,4-benzenediol, 2-tert-Butyl-1,4-dihydroxybenzene, 2-tert-Butyl-1,4-hydroquinone, 2-tert-Butyl-4-hydroxyphenol, 2-tert-Butylhydroquinone, Banox 20BA, Grindox 204, Grindox 450, Keniox 20, MC-TB 20, MTBHQ, Mono-tert-butylhydroquinone, Mono-tertiarybutylhydroquinone, NSC 4972, PI 172, Stearer TBH, TBHQ, Tenox 20, Tenox TBHQ, t-Butylhydroquinone, t-Butylhydroquinone, tert-Butyl-1,4-benzenediol, tert-Butyl-1,4-dihydroxybenzene, tert-Butyldihydroxybenzene, tert-Butylhydroquinone
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)c1cc(O)ccc1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H14O2/c1-10(2,3)8-6-7(11)4-5-9(8)12/h4-6,11-12H,1-3H3
Formula Weight 166.0994
Chemical Name or Material tert-Butylhydroquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.