missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit

Product Code. 10347121 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 10347121

Brand: Thermo Scientific Acros 349842500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2372-21-6 | C8H16O4 | 176.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2372-21-6
Molecular Formula C8H16O4
Linear Formula (CH3)2CHOCO3C(CH3)3
MDL Number MFCD00126878
Synonym tert-butoxy isopropyl carbonate, carbonoperoxoic acid, oo-1,1-dimethylethyl o-1-methylethyl ester, o,o-tert-butyl isopropyl monoperoxycarbonate, peroxycarbonic acid, oo-tert-butyl isopropyl ester, 2-methyl-2-propan-2-yloxy carbonyl peroxy propane, carbonic acid tert-butoxy isopropyl ester, oo-tert-butyl=o-isopropyl=peroxycarbonate, oo-t-butyl o-isopropyl monoperoxycarbonate, tert-butylperoxy isopropyl carbonate solution, 2-isopropoxycarbonyl peroxy-2-methylpropane
InChI Key KDGNCLDCOVTOCS-UHFFFAOYSA-N
SMILES CC(C)OC(=O)OOC(C)(C)C
IUPAC Name (2-methylpropan-2-yl)oxy propan-2-yl carbonate
Molecular Weight (g/mol) 176.212
PubChem CID 75406
Formula Weight 176.21
Quantity 250 mL
Chemical Name or Material tert-Butylperoxy isopropyl carbonate, 75% solution in Aromatic Free Mineral Spirit
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H242-Heating may cause a fire.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl peroxyisopropyl carbonate, Naphtha (petroleum) hydrotreated heavy

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.