missing translation for 'onlineSavingsMsg'
Learn More

Melford TES Free Acid

Product Code. 30018842
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018842 100 g 100g
30018843 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018842 Supplier Melford Supplier No. T31022100.0
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Substance Name: 2-Tris(hydroxymethyl)-methyl-2-amino 1-ethanesulfonic acid

CAS Number: 7365-44-8

Molecular Formula: C6H15NO6S

Molecular Weight: 229.25 g/mol

Specifications

Chemical Name or Material TES, Free Acid
CAS 7365-44-8
Quantity 100 g
Molecular Formula C6H15NO6S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.