missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford TBE 10X Solution
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
Description
TBE is a common buffer used in gel electrophoresis.
TBE is a common buffer used in gel electrophoresis.
Supplied as a 1X concentrated solution containing:
89mM Tris Base
89mM Boric Acid
2mM EDTA disodium salt
pH (1X solution 25°:C): 8.3+0.1
Specifications
Specifications
| CAS | 77-86-1, 10043-35-3, 6381-92-6, 7732-18-5 |
| Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction