Learn More
Invitrogen™ Tesnatilimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558359
 View more versions of this product
Product Code. 30283609
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tesnatilimab is an anti-CD38 monoclonal antibody that targets and binds to the CD38 protein on the surface of certain cells, leading to cell death through mechanisms such as antibody-dependent cellular cytotoxicity (ADCC) and complement-dependent cytotoxicity (CDC).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tesnatilimab
Recombinant Monoclonal
Unconjugated
D12S2489E; NKG2D
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized