missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Test Tube Stand

Product Code. 10037412 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
Up to 6 Eppendorf Vessels (1.5 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10037412 Up to 6 Eppendorf Vessels (1.5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10037412 Supplier Heidolph™ Supplier No. 5490400000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For use with Reax top and Reax control

For up to 6 Eppendorf vessels (1.5 mL)
TRUSTED_SUSTAINABILITY

Specifications

Holds Up to 6 Eppendorf Vessels (1.5 mL)
Tube Capacity 1.5 mL
Product Type Test Tube Stand
Color Blue
For Use With (Equipment) Reax top and Reax control
No. of Positions 6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.