missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Testis Tissue Slides (Adult Normal)- Paraffin

Product Code. 18098253 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18098253 5 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

This product is for research use only and is not approved for use in humans or in clinical diagnosis

Product Code. 18098253 Supplier R&D Systems™ Supplier No. NBP275940
  • ONLINE EXCLUSIVE PRICE
    356.04 €
    295.05 € / Pack of 5
    Save 60.99 €
    17% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is for research use only and is not approved for use in humans or in clinical diagnosis

A Novus product, part of R&D Systems. Tissue was fixed in formalin immediately after excision, and embedded in paraffin

  • Tissue from 24 year old Male Testis in normal condition
  • Store at 4°C, Do not freeze
  • Slides are guaranteed for 3 months from date of receipt
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5
Specimen Type testis tissue
Tissue Condition Normal Adult
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.