missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET2 Antibody (CL6873) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76510JF646
This item is not returnable.
View return policy
Description
TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| TET2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 | |
| This antibody was developed using a synthetic peptide derived from Q6N021, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL6873 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54790 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction