missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromo (R) Fluorescein, Pure, Fisher Chemical™

Product Code. 11463733 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463733 50 g 50g
10724581 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463733 Supplier Fisher Chemical Supplier No. T/0470/47

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number 5040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym eosin yellowish, eosine, bromo acid, eosine yellowish, eosine g, bromoeosine, c.i. acid red 87, sodium eosine, sodium eosinate, eosine salt free
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Melting Point 300°C
Color Brown-Red
pH 7 to 10
Packaging Glass Jar
Quantity 50 g
Formula Weight 691.85g/mol
Physical Form Powder
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.