missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromobisphenol A, TRC

Product Code. 30423089
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423089 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423089 Supplier Toronto Research Chemicals Supplier No. TRCT2911051G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79-94-7
Molecular Formula C15 H12 Br4 O2
Quantity 1 g
Synonym Phenol, 4,4'-(1-methylethylidene)bis[2,6-dibromo-, Phenol, 4,4'-isopropylidenebis[2,6-dibromo- (6CI,7CI,8CI), 4,4'-(1-Methylethylidene)bis[2,6-dibromophenol], 2,2-Bis(3,5-dibromo-4-hydroxyphenyl)propane, 2,2-Bis(4-hydroxy-3,5-dibromophenyl)propane, 2,2',6,6'-Tetrabromobisphenol A, 3,3',5,5'-Tetrabromobisphenol A, 3,5,3',5'-Tetrabromobisphenol A, 4,4'-Isopropylidenebis[2,6-dibromophenol], BA 59, BA 59BP, BA 59P, Bromdian, CP 2000, FCP 2010, FG 2000, FR 1524, Fire Guard 2000, Firemaster BP 4A, Flame Cut 120G, Flame Cut 120R, GLCBA 59P, NSC 59775, PB 100, RB 100, Saytex CP 2000, Saytex RB 100, Saytex RB 100PC, T 0032, TBBPA, Tetrabromobisphenol A, Tetrabromodian, Tetrabromodiphenylolpropane
InChI Formula InChI=1S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3
SMILES CC(C)(c1cc(Br)c(O)c(Br)c1)c2cc(Br)c(O)c(Br)c2
IUPAC Name 2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
Molecular Weight (g/mol) 543.87
Formula Weight 539.7571
Percent Purity >95%
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.