missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabromocatechol, 96%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
Tetrabromocatechol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrabromocatechol is used as a pharmaceutical intermediate.
Solubility
It is partly miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
Tetrabromocatechol is used as a pharmaceutical intermediate.
Solubility
It is partly miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 189°C to 193°C |
| Quantity | 5 g |
| Beilstein | 2051548 |
| Solubility Information | It is partly miscible with water. |
| Formula Weight | 425.7 |
| Percent Purity | 96% |
| Chemical Name or Material | Tetrabromocatechol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction