missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade
GREENER_CHOICE

Product Code. 10588322 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10588322 500 g Plastic bottle 500g
10055680 100 g Plastic bottle 100g
10023643 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10588322 Supplier Thermo Scientific Supplier No. 420125000
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.15 max. at 420nm
Color Undesignated
Linear Formula (CH3(CH2)3)4NOH
Beilstein 04, II, 634
Concentration 0.37 to 0.43M
Fieser 05,645; 11,500
Density 0.85
Solubility Information Solubility in water: soluble.
IUPAC Name tetrabutylazanium;hydroxide
Formula Weight 259.47
Grade HPLC
Packaging Plastic bottle
Quantity 500 g
Flash Point >110°C
Physical Form Solution
Chemical Name or Material Tetrabutylammonium hydroxide titrant, 0.4M in Water
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, 0.4M in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.