missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline hydrochloride, 96%

Product Code. 11489367
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11489367 100 g 100g
11479367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11489367 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B21408.22
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria. | CAS: 64-75-5 | C22H25ClN2O8 | 480.898 g/mol

Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria

Applications

  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Tekniske data

Type Tetracycline Hydrochloride
Melting Point 220°C to 223°C
Quantity 100 g
Assay Percent Range 0.98
Optical Rotation −247° (c=1, 0.1M HCl)
Sensitivity Air and light sensitive
Merck Index 14,9196
Solubility Information Soluble in water,dimethyl sulfoxide,methanol and ethanol. Insoluble in ether and hydrocarbons.
Formula Weight 480.91
Percent Purity 96%
Chemical Name or Material Tetracycline hydrochloride
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.