missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline hydrochloride, 96%, Thermo Scientific™

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.

47.80 € - 156.00 €

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479367
SDS View Documents
Thermo Scientific Alfa Aesar
B21408.14
25g
ONLINE EXCLUSIVE PRICE
48.80 €
47.80 € / 25g
Save 1.00 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11489367
SDS View Documents
Thermo Scientific Alfa Aesar
B21408.22
100g
ONLINE EXCLUSIVE PRICE
159.00 €
156.00 € / 100g
Save 3.00 €
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Tetracycline hydrochloride is the hydrochloride salt of
the antibiotic tetracycline
• Tetracycline blocks amino acyl-tRNA binding during
bacterial protein synthesis
• Tetracycline is a broad-spectrum naphthacene antibiotic that
inhibits growth of both gram positive and gram negative bacteria
Applications
• Tetracycline may be used as a selection agent in cell
culture
• It may also be used to activate inducible gene
expression in engineered cells
Specifications

Chemical Identifiers

64-75-5
480.898
HTXDZWDXSWLLLW-FMZCEJRJSA-N
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
C22H25ClN2O8
MFCD00078142
129628373
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

64-75-5
Air and light sensitive
MFCD00078142
Soluble in water,dimethyl sulfoxide,methanol and ethanol. Insoluble in ether and hydrocarbons.
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
480.898
480.91
Tetracycline hydrochloride
0.98
C22H25ClN2O8
14,9196
HTXDZWDXSWLLLW-FMZCEJRJSA-N
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
129628373
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H319

P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c

H319-H361d

missing translation for 'einecsNumber' : 200-593-8

missing translation for 'rtecsNumber' : QI9100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Tetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Tetracycline hydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.