missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetracycline hydrochloride, can be used as secondary standard

Product Code. 10626784 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Unit Size:
2.5g
500mg
Product Code. Quantity Packaging unitSize
10626784 2.5 g Glass Bottle 2.5g
10695272 500 mg Glass Bottle 500mg
2 options
This item is not returnable. View return policy

Product Code. 10626784

Brand: Thermo Scientific Chemicals 233100025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria

Applications

  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Type Tetracycline Hydrochloride
Physical Form Powder
Melting Point 223.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 252.00
Color Yellow
Quantity 2.5 g
Beilstein 14, IV, 2625
Merck Index 15, 9341
Solubility Information Solubility in water: 20g/L. Other solubilities: soluble in ethanol: 5g/L, insoluble in ether, hydrocarbons and acetone
Formula Weight 480.9
Percent Purity ≥95 (anhydrous and solvent free), 80 wt% (as is, calculated)
Chemical Name or Material Tetracycline hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.