missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride, Fisher BioReagents

Product Code. 10460264
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10460264 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10460264 Supplier Fisher BioReagents Supplier No. BP912100
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Tetracycline blocks protein synthesis and is used to select tetracycline-resistant recombinant plasmids.
TRUSTED_SUSTAINABILITY

Specifications

Type Tetracycline Hydrochloride
Physical Form Solid
CAS 64-75-5
pH 2.5
Melting Point 223°C
Loss on Drying 2.0% max.
Packaging Poly Bottle
Potency > 900μg/mg
Specific Rotation -255 to -240°
Color Yellow
Quantity 100 g
Molecular Formula C22H25ClN2O8
Merck Index 15, 9341
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 480.898
PubChem CID 129628373
Formula Weight 480.9
Chemical Name or Material Tetracycline Hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.