missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline, MedChemExpress

Product Code. 16451356
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451356 200 mg 200mg
16432019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451356 Supplier Medchem Express Supplier No. HYA0107
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tetracycline is a broad-spectrum antibiotic with oral activity. Tetracycline exhibits activity against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas and rickettsiae. Tetracycline can be used for the research of infections.

Tetracycline is a broad-spectrum antibiotic with oral activity. Tetracycline exhibits activity against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas and rickettsiae. Tetracycline can be used for the research of infections.

Specifications

Chemical Name or Material Tetracycline
Physical Form Powder
CAS 60-54-8
Color Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C22H24N2O8
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (281.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@](C)(O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N
Molecular Weight (g/mol) 444.43
Formula Weight 444.43
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.