missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl ethylenediphosphonate, 98%

Product Code. 11452287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452287 1 g 1g
11462287 5 g 5g
11472287 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452287 Supplier Thermo Scientific Alfa Aesar Supplier No. A14206.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 995-32-4 | C10H24O6P2 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 995-32-4
Molecular Formula C10H24O6P2
Molecular Weight (g/mol) 302.24
MDL Number MFCD00015133
InChI Key RSQYXXACEZCDFS-UHFFFAOYSA-N
Synonym tetraethyl ethylenediphosphonate, tetraethyl ethylenebisphosphonate, ethylenediphosphonic acid tetraethyl ester, tetraethyl ethane-1,2-diylbis phosphonate, 1,2-bis diethoxyphosphoryl ethane, 1,2-bis diethyl-phosphonato ethane, phosphonic acid, 1,2-ethanediylbis-, tetraethyl ester, diethyl 2-diethoxyphosphoryl ethylphosphonate, phosphonic acid, ethylenedi-, tetraethyl ester, acmc-209scy
PubChem CID 70446
IUPAC Name 1,2-bis(diethoxyphosphoryl)ethane
SMILES CCOP(=O)(CCP(=O)(OCC)OCC)OCC

Specifications

Density 1.146
Boiling Point 153°C to 154°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.441
Quantity 1 g
Beilstein 1800096
Formula Weight 302.25
Percent Purity 98%
Chemical Name or Material Tetraethyl ethylenediphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.