missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroterephthalonitrile, 98%

Product Code. 15417858
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417858 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15417858 Supplier Thermo Scientific Alfa Aesar Supplier No. H61326.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1835-49-0 | C8F4N2 | 200.096 g/mol

Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1835-49-0
Melting Point 197°C to 199°C
Molecular Formula C8F4N2
MDL Number MFCD00001776
Quantity 25 g
UN Number UN3439
Synonym tetrafluoroterephthalonitrile, 2,3,5,6-tetrafluoroterephthalonitrile, diamond 2031, 2,3,5,6-tetrafluoro-1,4-benzenedicarbonitrile, 1,4-benzenedicarbonitrile, 2,3,5,6-tetrafluoro, 1,4-dicyanotetrafluorobenzene, terephthalonitrile, tetrafluoro, perfluoroterephthalonitrile, tetrafluoroterephthalodinitrile, tetrafluorobenzene-1,4-dicarbonitrile
Solubility Information Soluble in hot methanol.
InChI Key PCRSJGWFEMHHEW-UHFFFAOYSA-N
SMILES C(#N)C1=C(C(=C(C(=C1F)F)C#N)F)F
IUPAC Name 2,3,5,6-tetrafluorobenzene-1,4-dicarbonitrile
Molecular Weight (g/mol) 200.096
PubChem CID 15783
Formula Weight 200.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Tetrafluoroterephthalonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Tetrafluoroterephthalonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrafluorophthalonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.