missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro Curcumin, TRC

Product Code. 30413281
Change view
Click to view available options
Quantity:
5 mg
25 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413281 1 g 1g
30424051 5 mg -
30423881 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413281 Supplier Toronto Research Chemicals Supplier No. TRCT2933851G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36062-04-1
Molecular Formula C21 H24 O6
Molecular Weight (g/mol) 372.41
Synonym 3,5-Heptanedione, 1,7-bis(4-hydroxy-3-methoxyphenyl)-, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione, 1,7-Bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione, AS-KTC 004, HZIV 81-2, NSC 687845, Sabiwhite, Tetrahydrocurcumin, Tetrahydrodiferuloylmethane
IUPAC Name 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione
SMILES COc1cc(CCC(=O)CC(=O)CCc2ccc(O)c(OC)c2)ccc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H24O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,24-25H,3-4,7-8,13H2,1-2H3
Formula Weight 372.1573
Chemical Name or Material Tetrahydrocurcumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.