missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrocannabinol Antibody (TH2A) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
323.00 € - 1050.00 €
Specifications
| Antigen | Tetrahydrocannabinol |
|---|---|
| Clone | TH2A |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626987
|
Novus Biologicals
NBP3-07896 |
1 mg |
1050.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655038
|
Novus Biologicals
NBP3-07896-0.1mg |
0.1 mg |
323.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Nonspecies specific samples. It is validated for ELISA, ImmunoassaySpecifications
| Tetrahydrocannabinol | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Tetrahydrocannabinol (THC) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| TH2A | |
| ELISA, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title