missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrocurcumin, MedChemExpress

Product Code. 16471578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471578 5 mg 5mg
16479168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471578 Supplier Medchem Express Supplier No. HYN0893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tetrahydrocurcumin is a Curcuminoid found in turmeric (Curcuma longa) that is produced by the reduction of Curcumin. Tetrahydrocurcumin inhibit CYP2C9 and CYP3A4.

Tetrahydrocurcumin is a Curcuminoid found in turmeric (Curcuma longa) that is produced by the reduction of Curcumin. Tetrahydrocurcumin inhibit CYP2C9 and CYP3A4.

Specifications

Chemical Name or Material Tetrahydrocurcumin
CAS 36062-04-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H24O6
Quantity 5 mg
Synonym HZIV 81-2
Solubility Information DMSO : 100 mg/mL (268.52 mM; Need ultrasonic)
SMILES O=C(CC(CCC1=CC=C(O)C(OC)=C1)=O)CCC2=CC=C(O)C(OC)=C2
Molecular Weight (g/mol) 372.41
Formula Weight 372.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.