missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)nickel(0), 95%

Product Code. 10012950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10012950 1 g Glass bottle 1g
10048711 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10012950 Supplier Thermo Scientific Acros Supplier No. 223980010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15133-82-1 | C72H60NiP4 | 1107.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15133-82-1
Molecular Formula C72H60NiP4
Molecular Weight (g/mol) 1107.86
MDL Number MFCD00010011
InChI Key KFBKRCXOTTUAFS-UHFFFAOYSA-N
Synonym ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
PubChem CID 11979963
IUPAC Name nickel;triphenylphosphane
SMILES [Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 122.0°C to 124.0°C
Color Brown to Brown-Rust
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Ni
Quantity 1 g
Fieser 06,570; 07,357; 09,450; 12,467; 13,294; 16,317
Formula Weight 1107.89
Percent Purity 95%
Physical Form Fine Powder
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0), 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrakis(triphenylphosphine)nickel(0)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Flammable solid Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.