missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)nickel(0), Thermo Scientific Chemicals

Product Code. 15481247
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481247 250 mg 250mg
15471247 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481247 Supplier Thermo Scientific Alfa Aesar Supplier No. 032101.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15133-82-1 | Molecular Formula: C72H60NiP4 | Molecular Weight: 1107.86 g/mol

Tetrakis(triphenylphosphine)nickel(0) is a catalyst for coupling reactions of organic halides, organometallic compounds and also for the cyclooligomerization of cumulenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15133-82-1
Molecular Formula C72H60NiP4
Molecular Weight (g/mol) 1107.86
MDL Number MFCD00010011
InChI Key KFBKRCXOTTUAFS-UHFFFAOYSA-N
Synonym ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
PubChem CID 11979963
IUPAC Name nickel;triphenylphosphane
SMILES [Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 123°C to 128°C
Color Brown
Linear Formula Ni[P(C6H5)3]4
Quantity 250 mg
UN Number UN1325
Sensitivity Air & Moisture Sensitive
Formula Weight 1107.83
Physical Form Crystalline
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.