missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)platinum(0), 98%

Product Code. 10390984 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10390984

Brand: Thermo Scientific Acros 223960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14221-02-4 | C72H60P4Pt | 1244.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14221-02-4
Molecular Formula C72H60P4Pt
Molecular Weight (g/mol) 1244.25
MDL Number MFCD00010014
InChI Key SYKXNRFLNZUGAJ-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
PubChem CID 11979705
IUPAC Name platinum;triphenylphosphane
SMILES [Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 159.0°C to 160.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Pt
Quantity 5 g
Fieser 15,300; 16,98; 04,33; 05,34; 09,45; 10,33; 14,36; 15,30; 16,29; 17,32; 06,570; 07,357; 09,450; 12,467; 13,294; 16,317
Solubility Information Solubility in water: insoluble.
Formula Weight 1244.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetrakis(triphenylphosphine)platinum(0)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.