missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide, 25 wt% in methanol

Product Code. 10274850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10274850 100 g Plastic bottle 100g
10284850 500 g Plastic bottle 500g
11975441 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10274850 Supplier Thermo Scientific Acros Supplier No. 138351000
  • ONLINE EXCLUSIVE PRICE
    42.15 €
    40.90 € / 100g
    Save 1.25 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 75-59-2 | C4H13NO | 91.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 25 wt. % in Methanol
CAS 75-59-2, 67-56-1
Boiling Point 65.0°C
Melting Point -98.0°C
Molecular Formula C4H13NO
Linear Formula (CH3)4NOH
MDL Number MFCD00008280
Density 0.9000g/mL
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.154
PubChem CID 60966
Formula Weight 91.15
Packaging Plastic bottle
Specific Gravity 0.9
Quantity 100 g
Flash Point 6°C
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide, 25 wt% in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methyl alcohol, Tetramethylammonium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.