missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide, 25 wt% in methanol

Product Code. 11975441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11975441 2.5 kg Plastic Bottle 2.5kg
10274850 100 g Plastic Bottle 100g
10284850 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11975441 Supplier Thermo Scientific Acros Supplier No. 138350025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 75-59-2 | C4H13NO | 91.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 25 wt. % in Methanol
CAS 75-59-2, 67-56-1
Boiling Point 65.0°C
Melting Point -98.0°C
Molecular Formula C4H13NO
Linear Formula (CH3)4NOH
MDL Number MFCD00008280
UN Number 3286
Density 0.9000g/mL
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.154
PubChem CID 60966
Formula Weight 91.15
Packaging Plastic Bottle
Specific Gravity 0.9
Quantity 2.5 kg
Flash Point 6°C
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.