missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, 99%

Product Code. 10338770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338770 100 mL Glass bottle 100mL
10073230 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10338770 Supplier Thermo Scientific Acros Supplier No. 268861000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-76-3 | C4H12Si | 88.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-76-3
Molecular Formula C4H12Si
Molecular Weight (g/mol) 88.23
MDL Number MFCD00008274
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
PubChem CID 6396
ChEBI CHEBI:85361
IUPAC Name tetramethylsilane
SMILES C[Si](C)(C)C

Specifications

Melting Point -99.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 26.0°C to 28.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Si(CH3)4
Refractive Index 1.3585
Quantity 100 mL
Beilstein 04, 625
Merck Index 15, 9377
Specific Gravity 0.64
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 88.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetramethylsilane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.