missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium bromide, 99%

Product Code. 10308200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10308200 25 g Plastic bottle 25g
10133790 5 g Glass bottle 5g
10743265 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10308200 Supplier Thermo Scientific Acros Supplier No. 204020250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2751-90-8
Molecular Formula C24H20BrP
Molecular Weight (g/mol) 419.30
MDL Number MFCD00011915
InChI Key BRKFQVAOMSWFDU-UHFFFAOYSA-M
Synonym tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide
PubChem CID 2724163
IUPAC Name tetraphenylphosphanium;bromide
SMILES [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 295.0°C to 299.0°C
Color White
Density 260g/mL
Flash Point 260°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (C6H5)4PBr
Quantity 25 g
Beilstein 16, II, 373
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohols
Formula Weight 419.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium bromide
Show More Show Less
compliance-icons
Product Identifier
  • Tetraphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.