missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEV Cleavage Site Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
330.00 € - 560.00 €
Specifications
| Antigen | TEV Cleavage Site |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648505
|
Novus Biologicals
NBP2-37831-25ug |
25 μg |
330.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761404
|
Novus Biologicals
NBP2-37831 |
50 μg |
560.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEV Cleavage Site Polyclonal specifically detects TEV Cleavage Site in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TEV Cleavage Site | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BBS (pH 8.4) with No Preservative | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Non-species specific | |
| Peptide representing the cleavage site recognized by TEV enzyme | |
| Primary | |
| This product has been successfully used in Western blot applications. The immunogen is a peptide representing the cleavage site recognized by TEV enzyme. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title