missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX30 Antibody (OTI7C3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44910-30ul
This item is not returnable.
View return policy
Description
TEX30 Monoclonal antibody specifically detects TEX30 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| TEX30 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 93081 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| OTI7C3 | |
| Western Blot 1:500 | |
| C13orf27, chromosome 13 open reading frame 27, hypothetical protein LOC93081, RP11-484I6.3, testis expressed 30 | |
| Full length human recombinant protein of human TEX30(NP_620134) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction