Learn More
TFEB Antibody (CL14112) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26853JF669
Product Code. 30302951
  • 770.00 € / 0.10mL
Description
TFEB Monoclonal antibody specifically detects TFEB in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TFEB
Monoclonal
Janelia Fluor 669
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB
This antibody was developed using a synthetic peptide derived from P19484, with the exact immunogen sequence remaining proprietary.
0.1 mL
Autophagy
7942
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry
CL14112
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified