missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEC Monoclonal Antibody (4F11)

Description
TFEC Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | TFEC |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4F11 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Tfec |
| Gene Accession No. | O14948 |
| Gene Alias | BB107417; bHLHe34; Class E basic helix-loop-helix protein 34; hTFEC-L; mTFEC; rTFEC; TCFEC; TFEC; TFE-C; TFECL; TFEC-L; Transcription factor EC; Transcription factor EC-like |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?