Learn More
Invitrogen™ TFPI2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA595444
Product Code. 16314715
  • 520.00 € / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse spleen tissue, HELA whole cell, human placenta tissue, MCF-7 whole cell. IHC: human placenta tissue. Flow: U87 cell, A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TFPI2
Polyclonal
Unconjugated
TFPI2
AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2
Rabbit
Affinity chromatography
RUO
21789, 7980
-20°C
Lyophilized
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
500 μg/mL
PBS with 4mg trehalose and 0.05mg sodium azide
O35536, P48307
TFPI2
A synthetic peptide corresponding to a sequence at the N-terminus of human TFPI2 (70-105aa EGNANNFYTWEACDDACWRIEKVPKVCRLQVSVDDQ).
100 μg
Primary
Human, Mouse