missing translation for 'onlineSavingsMsg'
Learn More

TG 100801, MedChemExpress

Product Code. 16463618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463618 10 mg 10mg
16426347 5 mg 5mg
16400017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463618 Supplier Medchem Express Supplier No. HY10186
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TG 100801 is a prodrug that generates TG 100572 by de-esterification in development to treat age-related macular degeneration. TG 100572 is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases; has IC50s of 2, 7, 2, 16, 13, 5, 0.5, 6, 0.1, 0.4, 1, 0.2 for VEGFR1, VEGFR2, FGFR1, FGFR2, PDGFRβ, Fgr, Fyn, Hck, Lck, Lyn, Src, Yes, respectively.

TG 100801 is a prodrug that generates TG 100572 by de-esterification in development to treat age-related macular degeneration. TG 100572 is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases

Specifications

Chemical Name or Material TG 100801
CAS 867331-82-6
Color Red
Purity Grade Notes Research
Molecular Formula C33H30ClN5O3
Quantity 10 mg
Solubility Information DMSO : 5.56 mg/mL (9.58 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1)OC2=CC=C(Cl)C(C3=CC(C)=C(N=C(NC(C=C4)=CC=C4OCCN5CCCC5)N=N6)C6=C3)=C2
Molecular Weight (g/mol) 580.08
Formula Weight 580.08
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.