Learn More
TGF-beta 1, 2, 3 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB1835R-100
 View more versions of this product
Product Code. 18760638
  • 503.00 € / 100µg
Description
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGF beta 1/2/3
Monoclonal
Unconjugated
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from cell culture supernatant
RUO
7040
Multi-species
IgG1
Neutralization
1D11R
Neutralization 0.25-1.25 ug/mL
ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Bovine bone-derived TGF-beta 1 and TGF-beta 2
100 μg
Primary
Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.