missing translation for 'onlineSavingsMsg'
Learn More

TGF-beta 1, 2, 3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18764938
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18764938 25 μg 25µg
18742648 100 μg 100µg
18780307 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18764938 Supplier R&D Systems Supplier No. MAB1835SP
  • ONLINE EXCLUSIVE PRICE
    245.00 €
    231.00 € / 25µg
    Save 14.00 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 143 publications

TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Bovine∣Canine∣Chicken∣Hamster∣Human∣Mink∣Mouse∣Multi-species∣Opossum∣Pig∣Rabbit∣Rat∣Rhesus Macaque∣Transgenic Mouse samples. It is validated for Activation∣Agonist Activity∣Binding Inhibition∣Bioassay∣Blocking∣Co-IP∣Depletion Assay∣Dot Blot∣ELISA Capture∣ELISA Capture (Matched Antibody Pair)∣ELISA Development∣ELISA Development (Capture)∣Flow Cytometry∣Functional Assay∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣In vivo Assay∣Luminex Development∣Neutralization∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF beta 1/2/3
Applications Activation, Agonist Activity, Binding Inhibition Assay, Bioassay, Blocking Assay, Co-immunoprecipitation (Co-IP), Depletion Assay, Dot Blot, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair), ELISA Development, ELISA Development (Capture), Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, Luminex, Neutralization, Western Blot
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Binding Inhibition, Neutralization 0.25-1.25 μg/mL, ELISA Capture (Matched Antibody Pair) 2-8 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Bovine bone-derived TGF-beta 1 and TGF-beta 2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. No cross-reactivity was observed with recombinant LAP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Bovine, Canine, Chicken, Hamster, Human, Mink, Mouse, Multi-species, Opossum, Pig, Rabbit, Rat, Rhesus Macaque, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.