Learn More
TGFA & ADAM17 Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DI0043.1Set
This item is not returnable.
View return policy
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TGFA protein, and the other against the ADAM17 protein for use in in situ Proximity Ligation Assay.Specifications
| TGFA | |
| Unconjugated | |
| TFGA | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. TGFA rabbit purified polyclonal antibody (20 ug) 2. ADAM17 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| transforming growth factor, alpha | |
| TGFA | |
| RUO | |
| 7039/6868 | |
| Human | |
| Ab Pair |