missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGR5/GPBAR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
254.00 € - 624.00 €
Specifications
| Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, KnockDown Validated |
|---|---|
| Classification | Polyclonal |
| Target Species | Human, Rat |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Isotype | IgG |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18730094
|
Novus Biologicals
NBP2-23669SS |
254.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18774993
|
Novus Biologicals
NBP2-23669 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGR5/GPBAR1 Polyclonal specifically detects TGR5/GPBAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, KnockDown Validated | |
| Human, Rat | |
| IgG | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Whole antisera with 0.05% Sodium Azide | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title