missing translation for 'onlineSavingsMsg'
Learn More

TH1L Mouse anti-Human, Unconjugated, Clone: 1A10G2, Proteintech

Product Code. 16849975 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16849975 150 μL 150µL
16859975 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16849975 Supplier Proteintech Supplier No. 673801IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TH1L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TH1L. This antibody recognizes human, mouse, rat antigen. The TH1L antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Antigen TH1L
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1A10G2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 51497
Target Species Human, Rat, Mouse
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.