missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THADA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93002-0.1ml
This item is not returnable.
View return policy
Description
THADA Polyclonal antibody specifically detects THADA in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| THADA | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100 | |
| death receptor-interacting protein, FLJ21877, FLJ44016, FLJ44876, gene inducing thyroid adenomas protein, KIAA1767, thyroid adenoma associated, thyroid adenoma-associated protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1684-1953 of human THADA (NP_071348.3). QLVILSCEDHLPTESRLAVVEVLTSTTPLFLTNPHPILELQDTLALWKCVLTLLQSEEQAVRDAATETVTTAMSQENTCQSTEFAFCQVDASIALALALAVLCDLLQQWDQLAPGLPILLGWLLGESDDLVACVESMHQVEEDYLFEKAEVNFWAETLIFVKYLCKHLFCLLSKSGWRPPSPEMLCHLQRMVSEQCHLLSQFFRELPPAAEFVKTVEFTRLRIQEERTLACLRLLAFLEGKEGEDTLVLSVWDSYAESRQLTLPRTEAAC | |
| 0.1 mL | |
| Cell Biology | |
| 63892 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction