missing translation for 'onlineSavingsMsg'
Learn More

Thallium for AAS 1g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12973295 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12973295 500 mL Bottle 500mL
12963295 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12973295 Supplier Fisher Chemical Supplier No. B 161 500
  • ONLINE EXCLUSIVE PRICE
    69.25 €
    53.96 € / 500mL
    Save 15.29 €
    22% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HNO3
Quantity 500 mL
Chemical Name or Material Thallium
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Thallium (Tl) 1000mg/l in HNO3 2%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.