missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, MedChemExpress

Product Code. 16471078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471078 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471078 Supplier Medchem Express Supplier No. HYB0809
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Theophylline (1,3-Dimethylxanthine) is a potent phosphodiesterase (PDE) inhibitor, adenosine receptor antagonist, and histone deacetylase (HDAC) activator.

Theophylline (1,3-Dimethylxanthine) inhibits PDE3 activity to relax airway smooth muscle. Theophylline (1,3-Dimethylxanthine) has anti-inflammatory activity by increase IL-10 and inhibit NF-κB into the nucleus. Theophylline (1,3-Dimethylxanthine) induces apoptosis. Theophylline (1,3-Dimethylxanthine) can be used for asthma and chronic obstructive pulmonary disease (COPD) research.

Specifications

Chemical Name or Material Theophylline
Physical Form Solid
CAS 58-55-9
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C7H8N4O2
For Use With (Application) Cancer-programmed cell death
Synonym 1,3-Dimethylxanthine Theo-24
Solubility Information DMSO : 11.11 mg/mL (61.67 mM; Need ultrasonic) ∣H2O : 5 mg/mL (27.75 mM; Need ultrasonic)
SMILES O=C(N1C)N(C)C2=C(N=CN2)C1=O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.