missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Acclaim™ PepMap™ Nano Series™ C18 Peptide Analysis HPLC Column, 3 μm, 1 mm x 50 mm
Shop All Thermo Scientific ProductsDescription
- High resolution in protein identification, biomarker discovery and systems biology
- Particular loadability allows hghest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
- The Acclaim PepMap nano LC columns offer lengths targeting fast or high resolution peptide separation
Specifications
Specifications
| Column Format | Capillary Column |
| Diameter (Metric) | 1 mm |
| Length (Metric) | 50 mm |
| Particle Size | 3 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| Surface Area | 450 m²/g |
| USP Type | L1 |
| Stationary Phase | C18 |
| Product Line | nanoLC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?