missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ AX Javelin™ Guard Columns

Product Code. 11533301 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
20 mm
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Unit Size
11533301 2.1 mm 20 mm Pack of 4
11513301 2.1 mm 10 mm Pack of 4
11523301 4 mm 10 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11533301 Supplier Thermo Scientific™ Supplier No. 73105022106
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offer convenient, easy-to-use protection for analytical HPLC columns

  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of BioBasic AX Solid Phase
  • Weak anion exchange phase for multiple charged species
  • 300Å pore size for enhanced protein and peptide separations
  • Suitable for HILIC retention and separation of highly polar molecules
  • Superb stability under demanding pH conditions

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase AX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.