missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Seal Diamond Heat Sealing Film

Product Code. 11540314
Change view
Click to view available options
Format:
Roll
Sheets
Unit Size:
370m
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
11540314 Sheets Pack of 100
11982361 Roll 370m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11540314 Supplier Thermo Scientific™ Supplier No. AB0812
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    171.00 € / Pack of 100
    Save 39.00 €
    18% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clear polymer heat seal is ideal for fluorescence and colorimetric applications

  • Suitable for PCR in screw-down or clip-down heated lid thermal cyclers
  • Good optical clarity, ideal for QPCR and other fluorescence applications
  • Peelable
  • Some solvent resistance
  • Seal integrity range: -80°C to +120°C

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description Diamond Clear Seal, Polmer, 85 x 135mm
Material Polyethylene
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
Dimensions (L x W) 80 x 125 mm
For Use With (Application) For ALPS 25 Automated Laboratory Plate Sealer
Format Sheets
Product Type Diamond Clear Seal
Product Line Abgene
Adhesive Polypropylene
Certified RNase/DNase-free No
Pierceable No
Temperature Range (Metric) -80°C to +120°C
Well Shape All well shapes
Plate Compatibility Polypropylene, Polyethylene, Cyclic Olefin Copolymer, Polystyrene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.